PURUS™ Frames for EnviroTack™ Contamination Control Mats
Designed to be used with the PURUS™ EnviroTack™ mat to create a system that is easy to use in any critical area where direct adhesion of mats to a flooring surface is not allowed.
$125.39 - $257.95
Specifications
Color White
View More Specs
Products 5
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity  
19-125-100
View Documents
PURUS™
PF203801W00
508 x 965 mm
Each for $125.39
19-125-102
View Documents
PURUS™
PF263801W00
26 x 38 in. (66 x 96.52 cm)
Each for $194.54
19-125-101
View Documents
PURUS™
PF204701W00
508 x 1194 mm
Each for $192.31
19-125-103
View Documents
PURUS™
PF284701W00
711 x 1194 mm
Each for $200.38
19-125-105
View Documents
PURUS™
PF384701W00
965 x 1194 mm
Each for $257.95
Description
Description
  • Easily moved for cleaning
  • Non-skid backing ensures frame stays in place
  • Fits all PURUS™ EnviroTack™ mats
  • Strong and Durable
  • Non-Slip made from Polymer Coated polyester textile
  • Naturally antimicrobial, mold and mildew resistant
  • Customization Available: Please call for details
Specifications
Specifications
White
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'