missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences Wheaton™ PVC-Coated Wire Rack for 60mL BOD Bottles

This rugged wire rack holds up to (20) 60mL bottles for drying or storage.

Supplier:  DWK Life Sciences 227729

Catalog No. 36-102-5185


Explore available promotions

Description

Description

  • Manufactured from rugged PVC coated wire
  • Coating is corrosion resistant
  • Racks interlock to connect stacked racks for safe transport
  • Made in the United States
Specifications

Specifications

Bottle Rack
Bottles (20 x 60 mL)
Yes, Interlocking
1 Each
Corrosion-resistant rack
PVC-coated Wire
BOD bottles
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions