Learn More
Eisco™ PVC Density Set
The PVC Density Set is a classic experiment for young minds to demonstrate that density does not change, regardless of volume and shape.
Supplier: Eisco™ PH0113G
Description
- 12 Rods varying in length from 1.5 in. to 2.625 in.
- To determine the density of the material the student plots mass versus volume and the slope therefore is the density (mass per unit volume).
- Students will need to do all measurements themselves so a balance and ruler are required.
- Base is made from a durable wood coated for years of use.
- Machined from top quality raw materials to produce accurate results and durability of decades of use.
Spécifications
| 12 Pk. | |
| Cylinder Set | |
| Wooden Holder | |
| PVC |