missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyrazine-2-carboxamidoxime, 97%, Thermo Scientific™
$76.53 - $76.53
Chemical Identifiers
CAS | 51285-05-3 |
---|---|
Molecular Formula | C5H6N4O |
Molecular Weight (g/mol) | 138.13 |
MDL Number | MFCD02258397 |
InChI Key | IXCAPURAZXLMLQ-UHFFFAOYSA-N |
Synonym | pyrazine-2-carboxamide oxime, nitroso 1h-pyrazin-2-ylidene methanamine, n'-hydroxy-2-pyrazinecarboximidamide, n-hydroxy-pyrazine-2-carboxamidine, n-hydroxypyrazine-2-carboximidamide, n'-hydroxypyrazine-2-carboximidamide, z-n'-hydroxypyrazine-2-carboximidamide, qzhjcrjemifnnb-uhfffaoysa-n, n-hydroxy-2-pyrazinecarboximidamide, n'-hydroxypyrazin-2-carboximidamide |
PubChem CID | 6762607 |
IUPAC Name | nitroso(1H-pyrazin-2-ylidene)methanamine |
SMILES | C1=CN=CC(=C(N)N=O)N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5483403
|
Thermo Scientific Chemicals
H5483403 |
1 g |
Each for $76.53
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
51285-05-3 | |
138.13 | |
IXCAPURAZXLMLQ-UHFFFAOYSA-N | |
6762607 | |
C1=CN=CC(=C(N)N=O)N1 |
C5H6N4O | |
MFCD02258397 | |
pyrazine-2-carboxamide oxime, nitroso 1h-pyrazin-2-ylidene methanamine, n'-hydroxy-2-pyrazinecarboximidamide, n-hydroxy-pyrazine-2-carboxamidine, n-hydroxypyrazine-2-carboximidamide, n'-hydroxypyrazine-2-carboximidamide, z-n'-hydroxypyrazine-2-carboximidamide, qzhjcrjemifnnb-uhfffaoysa-n, n-hydroxy-2-pyrazinecarboximidamide, n'-hydroxypyrazin-2-carboximidamide | |
nitroso(1H-pyrazin-2-ylidene)methanamine |
Specifications
51285-05-3 | |
MFCD02258397 | |
pyrazine-2-carboxamide oxime, nitroso 1h-pyrazin-2-ylidene methanamine, n'-hydroxy-2-pyrazinecarboximidamide, n-hydroxy-pyrazine-2-carboxamidine, n-hydroxypyrazine-2-carboximidamide, n'-hydroxypyrazine-2-carboximidamide, z-n'-hydroxypyrazine-2-carboximidamide, qzhjcrjemifnnb-uhfffaoysa-n, n-hydroxy-2-pyrazinecarboximidamide, n'-hydroxypyrazin-2-carboximidamide | |
C1=CN=CC(=C(N)N=O)N1 | |
138.13 | |
138.13 | |
Pyrazine-2-carboxamidoxime |
C5H6N4O | |
1 g | |
IXCAPURAZXLMLQ-UHFFFAOYSA-N | |
nitroso(1H-pyrazin-2-ylidene)methanamine | |
6762607 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures