missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrene, 98%, Thermo Scientific Chemicals

$99.92 - $270.22

Chemical Identifiers

CAS 129-00-0
Molecular Formula C16H10
Molecular Weight (g/mol) 202.256
MDL Number MFCD00004136
InChI Key BBEAQIROQSPTKN-UHFFFAOYSA-N
Synonym benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene
PubChem CID 31423
ChEBI CHEBI:39106
IUPAC Name pyrene
SMILES C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0816214
View Documents
Thermo Scientific Chemicals
L0816214
25 g
Each for $99.92
 
AAL0816222
View Documents
Thermo Scientific Chemicals
L0816222
100 g
Each for $270.22
 
Description

Description

Pyrene and its derivatives are used commercially to make dyes and dye precursors, for example pyranine and naphthalene-1,4,5,8-tetracarboxylic acid. It is used as a probe to determine solvent environments and fluorescence spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrene and its derivatives are used commercially to make dyes and dye precursors, for example pyranine and naphthalene-1,4,5,8-tetracarboxylic acid. It is used as a probe to determine solvent environments and fluorescence spectroscopy.

Solubility
Soluble in toluene. Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
Specifications

Chemical Identifiers

129-00-0
202.256
BBEAQIROQSPTKN-UHFFFAOYSA-N
31423
pyrene
C16H10
MFCD00004136
benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene
CHEBI:39106
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2

Specifications

129-00-0
1.271
208°C (406°F)
MFCD00004136
1307225
benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene
BBEAQIROQSPTKN-UHFFFAOYSA-N
pyrene
31423
202.26
Pyrene
148°C to 152°C
393°C
C16H10
25 g
14,7963
Soluble in toluene. Insoluble in water.
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
202.256
CHEBI:39106
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H332
Harmful if inhaled.

P260-P271-P304+P340-P312-P314-P501c

H336-H373

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 204-927-3

RTECSNumber : UR2450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only