missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyrene 97.0+%, TCI America™
$105.54 - $1256.05
Chemical Identifiers
CAS | 129-00-0 |
---|---|
Molecular Formula | C16H10 |
Molecular Weight (g/mol) | 202.256 |
MDL Number | MFCD00004136 |
InChI Key | BBEAQIROQSPTKN-UHFFFAOYSA-N |
Synonym | benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene |
PubChem CID | 31423 |
ChEBI | CHEBI:39106 |
IUPAC Name | pyrene |
SMILES | C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2 |
Chemical Identifiers
129-00-0 | |
202.256 | |
BBEAQIROQSPTKN-UHFFFAOYSA-N | |
31423 | |
pyrene |
C16H10 | |
MFCD00004136 | |
benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene | |
CHEBI:39106 | |
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2 |
Specifications
129-00-0 | |
404°C | |
MFCD00004136 | |
2811 | |
BBEAQIROQSPTKN-UHFFFAOYSA-N | |
pyrene | |
31423 | |
202.26 | |
Crystalline Powder |
150°C | |
C16H10 | |
25 g | |
benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene | |
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2 | |
202.256 | |
CHEBI:39106 | |
≥97.0% (GC) | |
Pyrene |
Safety and Handling
EINECSNumber : (4)-0782
RTECSNumber : UR2450000
TSCA : Yes