Corning™ PYREX™ 1L Replacement Flasks With Stopper
Flat bottom, side delivery tube
$585.00 - $660.41
Specifications
Material Borosilicate Glass
Includes Side delivery tube and Pyrex™ No. 27 standard taper stopper
No. per Case 1/Cs.
No. per Pack 1/Pk.
Standard Taper Joint 19/38
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
09-126-5B
View Documents
Corning™
33601LFO
1 L
Case for $660.41
09-126-5A
View Documents
Corning™
3360500FO
500 mL
Case for $585.00
Description
Description
This PYREX flat bottom flask has a standard taper PYREXstoppered neck and a side delivery tube, terminating in a standard taper joint. It is a replacement flask for the General Purpose Distillation Apparatus No. 3360. For replacement stoppers, see No. 7650. For replacement Graham condensers, see No. 2540-200.
Specifications
Specifications
Borosilicate Glass
1/Cs.
19/38
Side delivery tube and Pyrex™ No. 27 standard taper stopper
1/Pk.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'