Corning™ PYREX™ 3-Hole Mobile Phase Delivery Cap
Designed for use with Corning PYREX HPLC Reservoirs, Single Cavity (sold separately). See Cat. Nos. 13-645-095, 13-645-096, 13-645-097, or 13-645-098.
Supplier:  Corning™ 13953H
Catalog No. 13-645-099
Explore available promotions
Description
Description
  • Delivery cap top has three holes that have 0.25 in. - 28 threads with 0.125 O.D. PTFE tubing
  • Body is glass-filled PTFE
  • PET screw cap with GL-45 thread
  • Cap includes three 0.25 in. - 28 ETFE plugs and Viton O-ring for sealing to the reservoir
  • Integral pressure relief valve relieves reservoir pressure at 10 psi
Order Info
Designed for use with Corning PYREX HPLC Reservoirs, Single Cavity (sold separately). See Cat. Nos. 13-645-095, 13-645-096, 13-645-097, or 13-645-098.
Specifications
Specifications
PTFE
Three 0.25 in. -28 ethylene tetrafluoroethylene (ETFE) plugs and a fluorocarbon O-ring for sealing to the reservoir
1/Pk.
3-Hole Mobile Phase Delivery Cap
1/Cs.