Corning™ PYREX™ Buret with Locking Stopcocks Replacement Components
For use with Pyrex 25mL economy-grade burets.
$182.00 - $933.44
Products 3
Catalog Number Mfr. No. Capacity (Metric) Type Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Type Price Quantity  
03-700-15C
View Documents
Corning™
211625BO
25 mL Economy Buret
Case of 6 Each for $857.35
03-700-15D
View Documents
Corning™
211650BO
50 mL Economy Buret
Case of 6 Each for $933.44
03-700-15F
View Documents
Corning™
2116LSO
25 mL Stopcock
Case for $182.00
Description
Description
An economical buret designed to reduce replacement costs by providing replaceable components. These sturdy, durable burets have tooled tops and bottoms and white enamel graduations. The screw thread locking PTFE stopcock with ground tip is compatible with Cat. No. 2116 existing buret bodies. Simply replace the stopcock assembly to benefit from the improved design. The new replaceable stopcock assembly features a screw thread locking nut and collar which ensures that the stopcock cannot fall out in use. Cat. No. 2116-GTO replaceable tip has a ground end which ­provides a better grip and helps prevent the tip from falling out.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'