Corning™ PYREX™ Cloning Cylinders
Create identical cells by surrounding a single cell or group of cells
Supplier:  Corning™ 31668
 View more versions of this product
Catalog No. 09-552-21
Explore available promotions
Description
Description
Clone a single cell or group of cells by surrounding them with this glass cylinder. Dip the end of the cylinder into a sterile silicone grease before pressing to the bottom of a culture flask to create an ­isolated well.
Specifications
Specifications
8 mm
8 mm
0.31 in.
0.31 in.
Non-sterile
Borosilicate Glass
Cloning Cylinder
Product Suggestions
Product Suggestions