Learn More
Corning™ PYREX™ Connecting 3-Way Claisen Adapters with Parallel Side Arms
Three-way connecting tube with two outer and one inner standard taper 24/40 joints
Supplier: Corning™ 904024
Alert:
Because of variations in conditions, glassware cannot be guaranteed against breakage under vacuum or pressure. Adequate precautions should be taken to protect personnel who are doing such work.
Description
Cat. No. 9040-24 has outer standard taper joints at the upper end and on the parallel sidearm. With inner joint at the lower end. All joints on each tube are of the same size.
Cat. Nos. 9050-14 and 9050-19 have a three-way tube with two outer and one inner joint of the same size.
Specifications
Connecting Adapter | |
6.29 in. | |
24/40 Inner bottom, 24/40 outer top |
Borosilicate Glass (3.3) | |
16 cm |