missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ PYREX™ Cylindrical Jars With Handles

Mold-blown with substantial walls; ground flat open end

Supplier:  Corning™ 694327L

 View more versions of this product

Catalog No. 11-822B


Explore available promotions

Description

Description

These jars are made with recessed finger grips for convenience in handling. They are identical in size and shape to corresponding jars sizes of Cat. No. 6942. Open ends are finely ground. Do not use with heat, pressure, or vacuum applications.

Specifications

Specifications

932.6 oz., 26500 mL
932.6 oz.
Recessed Handle
1/Cs.
Round
Cylindrical Jar
305 mm
26,500 mL
12 in.
Glass
1/Pk.
457 mm
17.9 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions