Corning™ PYREX™ Disposable Rimless Flat-Bottom Glass Tubes
Ideal for plant tissue culture and general culture work
Supplier:  Corning™ 985025X
 View more versions of this product
Catalog No. 14-957-85B
Description
Description
These PYREX™ flat bottom reusable culture tubes are ideal for plant tissue culture and general culture work. The flat bottom facilitates use with agar slants. Walls and bottoms are of uniform thickness. The tubes are rimless for greater convenience in plugging and rack storage. Optional polypropylene two position closures (Nos. 26794-25 or 26795-25) can be opened for gas exchange or closed for humidified environment. Internal drip ring minimizes contamination.
Order Info
Supplied without closures; except Pyrex 14-957-85E, -F, -G, and -K caps (sold separately).
Specifications
Specifications
47 mL
Borosilicate Glass
For tissue culture work and general bacteriological use
No
No
Culture Tube
120 mm
No
None
Non-sterile
25 mm