PYREX™ Enlarging Glass Joint Adapters
For attaching apparatus with one standard taper joint size to equipment having larger size standard taper joint
Supplier:  PYREX™ 88202445
 View more versions of this product
Catalog No. 01-034F
Explore available promotions
Description
Description
With an outer standard taper joint at the top and a larger inner standard taper joint at the bottom.
Specifications
Specifications
For attaching apparatus with one joint size to equipment having larger size joint
115 mm
Tube Adapter
4.52 in.
Borosilicate Glass
45/50 Inner Bottom, 24/40 Outer Top