Corning™ PYREX™ Evaporator Traps, Standard Taper Joints
Used when foam is likely to be created and drawn into the condenser of rotary evaporator
$446.12 - $510.43
Specifications
Material Borosilicate Glass
Product Type Rotary Evaporator Trap
For Use With (Application) Used when foam is likely to be created and drawn into the condenser of rotary evaporator
For Use With (Equipment) Rotary evaporator
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (English) Diameter (English) Outer Standard Taper Joint Length (English) Price Quantity  
Catalog Number Mfr. No. Capacity (English) Diameter (English) Outer Standard Taper Joint Length (English) Price Quantity  
09-549-2A
View Documents Promotion Details
Corning™
400010019
3.4 oz. 2.4 in. Upper: 24/40, Lower: 19/22 6.3 in.
Case for $446.12
09-549-3A
View Documents Promotion Details
Corning™
400010024
3.4 oz. 2.4 in. Upper: 24/40, Lower: 24/40 7 in.
Case for $481.10
09-549-3B
View Documents Promotion Details
Corning™
400025024
8.45 oz. 3.1 in. Upper: 24/40, Lower: 24/40 7.8 in.
Case for $510.43
Description
Description
  • Round body with upper and lower stems
  • Lower stem, with standard taper inner joint at bottom, extends upward inside the body
  • Closed top of lower stem has two side openings
  • Upper stem has standard taper 24/40 outer joint
Specifications
Specifications
Borosilicate Glass
Rotary Evaporator Trap
Used when foam is likely to be created and drawn into the condenser of rotary evaporator
Rotary evaporator
SDS
missing translation for 'documents'
missing translation for 'documents'