Corning™ PYREX™ Powder Funnel
With short wide stems
$272.15 - $2101.30
Specifications
Material Borosilicate Glass
For Use With (Application) For transferring powders or filling bottles
View More Specs
Products 5
Catalog Number Mfr. No. Diameter (Metric) Stem Diameter (Metric)Top Length (Metric) Stem Height (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Stem Diameter (Metric)Top Length (Metric) Stem Height (Metric) Price Quantity  
10-346D
View Documents
Corning™
6220125
2.5 cm 12.5 cm 2.5 cm 13.2 cm
Case of 12 Each for $1,013.48
10-346A
View Documents
Corning™
622065
1.5 cm 6.5 cm 2.5 cm 8.3 cm
Pack of 12 Each for $485.61
Case of 2 Pack for $971.22
10-346B
View Documents
Corning™
622075
1.7 cm 7.5 cm 1.7 cm 9.4 cm
Pack of 12 Each for $458.16
Case of 2 Pack for $916.32
10-346C
View Documents
Corning™
6220100
2 cm 10 cm 2 cm 12 cm
Pack of 6 Each for $272.15
Case of 4 Pack for $1,088.60
10-346E
View Documents
Corning™
6220150
3 cm 15 cm 3 cm 15.7 cm
Case of 6 Each for $2,101.30
Description
Description
With short wide stems
These funnels have a short stem. For use in transferring powders or filling bottles.
Specifications
Specifications
Borosilicate Glass
For transferring powders or filling bottles
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'