Corning™ PYREX™ 60° Angle Fluted Funnel, Short Stem
Useful in clinical and hospital labs; can be used in flasks or tubes without racks
Supplier:  Corning™ 618065
 View more versions of this product
Catalog No. 10-329B
Explore available promotions
Description
Description
Supplied with short stems. Depressed inside flutings further decreases filtering time.
Specifications
Specifications
6.5 cm
Borosilicate Glass
60°
6.5 cm
0.8 cm
12.1 cm
Useful in clinical and hospital labs, can be used in flasks or tubes without racks
Fluted Filling Funnel