Corning™ PYREX™ Glass Wool
Strong, soft, pliable glass wool is ideal for general lab and industrial applications. Suitable for filtering.
Supplier: Corning™ C3950
Description
For general lab and industrial applications, this strong, soft, pliable glass wool is suitable for filtering.
This fiber glass is made from fine glass fibers with diameters controlled to approximately 0.008mm. This makes the glass pliable and easy to use. Manufactured from No. 9989 glass, this fiber glass contains minimal heavy metals and is suitable for use as a filtering medium. It is supplied in the form of roving approximately 2 inches in diameter by 22 ft. long and packaged in 0.23 kg (1â2 lb.) bags. This product is only sold in a case qty of 2.72 kg (6 lbs.).Order Info
Supplied in the form of roving approximately 2 in. diameter (5cm) and 22 ft. long (6.7m); wrapped in 1/2 lb. (226g) bags
Specifications
Each | |
For general laboratory and industrial use, suitable for filtering | |
0.008 mm |
Glass Wool | |
Soda-Lime Borosilicate Glass |