Learn More
Corning™ PYREX™ Heavy-Duty Griffin Beakers
For use under harsh conditions such as mechanized washing
$160.50 - $417.46
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
02-555-25A
|
Corning™
1003150 |
150 mL |
Pack of 12 Each for $260.39
Case of 4 Pack for $1,041.56
|
|
|||||
02-555-25B
![]() |
Corning™
1003250 |
250 mL |
Pack of 12 Each for $211.33
Case of 4 Pack for $845.32
|
![]() |
|||||
02-555-25C
|
Corning™
1003400 |
400 mL |
Pack of 12 Each for $280.07
Case of 4 Pack for $1,120.28
|
|
|||||
02-555-25D
|
Corning™
1003600 |
600 mL |
Pack of 6 Each for $160.50
Case of 6 Pack for $963.00
|
|
|||||
02-555-25F
|
Corning™
10031L |
1 L |
Pack of 6 Each for $306.28
Case of 4 Pack for $1,225.12
|
|
|||||
02-555-25H
|
Corning™
10032L |
2000 mL |
Pack of 4 Each for $417.46
Case of 2 Pack for $834.92
|
|
|||||
02-555-25K
|
Corning™
10034L |
4000 mL |
Each for $246.68
Case of 4 Each for $986.72
|
|
|||||
Description
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes
Measure liquids with sturdy, long-lasting, reinforced glass. Pyrex™ Borosilicate Glass Heavy-duty Griffin Beakers are available in a wide range of sizes.
Heavy duty beakers specifically designed to offer the best mechanical strength under harsh conditions, such as mechanized washing operations. For convenience, the 250 through 4000 mL beakers have a double graduated metric scale to indicate their approximate content. All sizes have an extra large marking spot.
Compliance
Designed to ASTM™ E 960 requirements.
Specifications
Borosilicate Glass | |
Beaker | |
Yes | |
Griffin |