missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ PYREX™ Microchemistry Kit Accessories - Round-bottom Reaction Flasks, Standard Taper Joint
Replacement part for PYREX Microchemistry Kits
$1368.67 - $1435.91
Specifications
Product Type | Reaction Flask |
---|---|
For Use With (Equipment) | For the 6949m series of Pyrex™ microchemistry kit |
Includes | Screw thread cap |
Material | Pyrex™ Glass |
Standard Taper Joint | 14/10 |
Description
This round bottom flask is a replacement part for the 6949M series of PYREX™ microchemistry kits.
Order Info
For 11-300-71 and 11-300-74 Microchemistry Kits
Specifications
Reaction Flask | |
Screw thread cap | |
14/10 |
For the 6949m series of Pyrex™ microchemistry kit | |
Pyrex™ Glass |