missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ PYREX™ Potter-Elvehjem Tissue Grinder with PTFE Pestle
Used in preparing small samples for pathological and biochemical studies
Supplier: Corning™ 7725T8
Alert:
If used with a motor-driven device, the attachment must contain some form of friction release clutch to prevent damage to the apparatus from pieces of connective tissue or muscle. The user should wear a protective glove of sufficient thickness to prevent injury in the event of failure of the glass components.
Description
Homogenization occurs as the sample and buffer are forced through the cylindrical portion of the mortar as the pestle is rotated downward. Mortar is unground and pestle contact surface is PTFE threaded onto a stainless steel shaft.
Caution: If used with a motor-driven device, the attachment must contain some form of friction release clutch to prevent damage to the apparatus. The user should wear a protective glove of sufficient thickness to prevent injury in the event of failure of the glass Âcomponents.
Order Info
Sold in a Case of 1.
Specifications
Non-sterile | |
8 mL | |
Potter-Elvehjem Tissue Grinder | |
Case of 1 | |
145 mm | |
Tooled mouth | |
0.27 oz., 8 mL |
0.27 oz. | |
Borosilicate Glass | |
Preparing small samples for pathological and biochemical studies | |
5.7 in. | |
0.7 in. | |
18 mm | |
PTFE |