Corning™ PYREX™ PTFE Product Standard Straight Bore Stopcock Plug Assembly
Replacement PTFE stopcock plug assemblies for straight bore PTFE stopcocks
Supplier: Corning™ 76816
Description
These are replacement Polytetrafluorethylene (PTFE) Product Standard stopcock plug assemblies with various diameter bores for Cat. No. 7282 PYREX straight bore PTFE stopcocks. For more information on PTFE stopcocks, please check the Care of Corning PYREX™ PTFE Stopcocks under Technical Literature.
Order Info
Case of 1
Specifications
Stopcock Plug Assembly, O-ring Size B | |
PTFE | |
1/Cs. |
6 mm | |
1/Pk. | |
No. 7282 Pyrex straight bore PTFE stopcocks |