Corning™ PYREX™ Reducing Glass Joint Adapters
For attaching apparatus with one standard taper joint size to equipment having a different size standard taper joint
Supplier:  Corning™ 88002924
 View more versions of this product
Catalog No. 01-035E
Explore available promotions
Description
Description
For attaching apparatus with one standard taper joint size to equipment having a different size standard taper joint
With an outer standard taper joint at the top and a smaller inner standard taper joint at the bottom.
Specifications
Specifications
For attaching apparatus with one joint size to equipment having a different size joint
105 mm
Reducing Adapter
4.13 in.
Borosilicate Glass
24/40 Inner Bottom, 29/42 Outer Top