Corning™ PYREX™ Reusable Class A Volumetric Pipets, Color-Coded
Borosilicate-glass with fire-polished top and tip
Supplier:  Corning™ 71002
 View more versions of this product
Catalog No. 13-651-1B
Explore available promotions
Description
Description
Borosilicate-glass with fire-polished top and tip
These pipets are manufactured to Class A capacity tolerances as indicated by ASTM E-542, ASTM E-694, and ASTM E-969. The rugged construction and colored enamel graduations and markings provide a durable pipet. Sizes 1 mL through 25 mL are color-coded.
Cat. No. 7100-50 new design allows more complete and efficient drainage. Do not pipet by mouth. We suggest using a mechanical pipetting device.
Certifications
NIST traceable
Compliance
Calibrated To Deliver with tolerances in accordance with ASTM™ E 542 and E 969
Specifications
Specifications
Autoclavable
To Deliver
A
Non-sterile
Round
Orange
33.3 cm
Reusable
Yes
ASTM E542, ASTM E969, certificate of identification and capacity, NIST traceable
Borosilicate Glass
13.11 in.
±0.006 mL
2 mL
Serialized Volumetric Pipet
Mechanical Pipetting Device
Product Suggestions
Product Suggestions