Learn More
Corning™ PYREX™ Reusable Class A Volumetric Pipets, Color-Coded
Borosilicate-glass with fire-polished top and tip
Supplier:  Corning™ 710025
 Afficher plus de versions de ce produit
Numéro de catalogue. 13-651-1N
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Borosilicate-glass with fire-polished top and tip
These pipets are manufactured to Class A capacity tolerances as indicated by ASTM E-542, ASTM E-694, and ASTM E-969. The rugged construction and colored enamel graduations and markings provide a durable pipet. Sizes 1 mL through 25 mL are color-coded.
Cat. No. 7100-50 new design allows more complete and efficient drainage. Do not pipet by mouth. We suggest using a mechanical pipetting device.
Certifications
NIST traceable
Compliance
Calibrated To Deliver with tolerances in accordance with ASTM™ E 542 and E 969
Spécifications
Spécifications
Autoclavable
To Deliver
A
Non-sterile
Round
Blue
46 cm
Reusable
Yes
ASTM E542, ASTM E969, certificate of identification and capacity, NIST traceable
Borosilicate Glass
18.11 in.
±0.03 mL
25 mL
Serialized Volumetric Pipet
Mechanical Pipetting Device
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS