Learn More
Corning™ PYREX™ Reusable Class B Mohr (Measuring) Pipets, TD, Color-Coded
Feature fire-polished top and tip
$377.30 - $709.59
Specifications
Certifications/Compliance ASTM E1293
Class B
Sterility Non-sterile
Graduated Yes
Tip Style Round
View More Specs
Products 5
Catalog Number Mfr. No. Volume (Metric) Graduations Color Tolerance Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Graduations Color Tolerance Price Quantity  
13-665-100F
View Documents
Corning™
706525
25 mL 0.1 mL White ±0.1 mL
Case of 12 Each for $709.59
Only null left
13-665-101A
View Documents
Corning™
70651
1 mL 0.1 mL Red ±0.02 mL
Case of 12 Each for $388.25
Only null left
13-665-101C
missing translation for 'viewDocuments'
Corning™
70652
2 mL 0.1 mL White ±0.02 mL
Case of 12 Each for $377.30
Only null left
13-665-101D
missing translation for 'viewDocuments'
Corning™
70655
5 mL 0.1 mL Blue ±0.04 mL
Case of 12 Each for $452.90
Only null left
13-665-101E
missing translation for 'viewDocuments'
Corning™
706510
10 mL 0.1 mL Orange ±0.06 mL
Case of 12 Each for $508.54
Only null left
Description
Description
Feature fire-polished top and tip
These pipets are color-coded by size for easy identification and sorting. Colored graduations are enameled onto the glass. The top end of 5 mL to 25 mL sizes is constricted (Mohr type). They are ­calibrated 'To Deliver' their total capacity without blow-out.
Do not pipet by mouth. We suggest using a mechanical pipetting device.
Reference: ASTM E-1293.
Compliance
Designed from ASTM™ E 1293.
Specifications
Specifications
ASTM E1293
Non-sterile
Round
Reusable
B
Yes
Graduated Pipet, Type 1