missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ PYREX™ Reusable Media Storage Bottles

Ideal for media, buffers, aqueous and nonaqueous solutions.

Supplier:  Corning™ 13955L

 View more versions of this product

Catalog No. 06-414-1F


Explore available promotions

Alert:

Bottles should not have caps tightened immediately after autoclaving, nor should they be used with bottle-top filter units or other applications involving vacuum pressure, as breakage may occur.


Description

Description

Ideal for media, buffers, aqueous and nonaqueous solutions

Heavy duty bottle which can be used for storage as well as mixing and sampling. Includes linerless, one-piece autoclavable polypropylene plug seal cap with drip free pouring rings. Optional red PBT caps and red ETFE pouring rings are available for dry heat sterilization (180°C). Glass bead indicates full capacity line on 100 mL through 2L size. Bottles have permanent white enamel graduations and marking spots. Color-coded polypropylene caps are available in a choice of orange, green, purple, red, and light gray.

Caution: Bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT have caps tightened immediately after autoclaving, as the vacuum resulting from cooling can cause breakage. A pouring ring is not included on 25 mL size. A vented cap is NOT recommended for use on the 5L and 10L 1395 series bottle.

Order Info

Optional red melamine caps and PTFE rings are available for applications requiring temperatures to 180°C (356°F).

Specifications

Specifications

169.07 oz., 5000 mL
169.07 oz.
Borosilicate Glass
1/Pk.
Non-sterile
500 to 4500 mL
Cylindrical
13.1 in.
GL 45
Plug Seal
5000 mL
7.3 in.
1/Cs.
1/Cs.
Orange
Wide
335 mm
Polypropylene
186 mm
Videos
SDS
Documents

Documents

Promotions

Promotions