missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ PYREX™ Squibb Separatory Funnel with Standard Taper Joing Stopcock and Stopper
Separatory funnel, used in liquid-liquid extractions to separate the components of a mixture into two immiscible solvent phases of different densities.
Supplier: Corning™ 64001L
Description
For stopper only, see Cat. No. 7650.
For stopcock only, see Cat. No. 7280.
Specifications
11.9 cm | |
Borosilicate Glass | |
6.5 cm | |
4 | |
Squibb Separatory Funnel |
1 L | |
39.2 cm | |
Each | |
27 |