Corning™ PYREX™ Squibb Separatory Funnel with Standard Taper Joing Stopcock and Stopper
Separatory funnel, used in liquid-liquid extractions to separate the components of a mixture into two immiscible solvent phases of different densities.
Supplier:  Corning™ 64004L
 View more versions of this product
Catalog No. 10-437G
Explore available promotions
Description
Description
For stopper only, see Cat. No. 7650.
For stopcock only, see Cat. No. 7280.
Specifications
Specifications
20.6 cm
Borosilicate Glass
6.5 cm
8
Squibb Separatory Funnel
4 L
62.9 cm
1/Cs.
38