Corning™ PYREX™ Thermometer Adapter
Component part for organic chemistry kit
Supplier:  Corning™ 882124
 View more versions of this product
Catalog No. 01-034-1C
Explore available promotions
Description
Description
One end with a standard taper ground joint and the other end tooled to accommodate a thermometer. The adapter is supplied complete with a rubber thermometer holder (Cat. No. 7715).
This tube is also a replacement part for all organic chemistry kit (Cat. No. 6949 through 6949K).
Specifications
Specifications
Replacement rubber thermometer holders
80 mm
Straight Adapter
Thermometers
Rubber thermometer holder
Borosilicate Glass
3.14 in.