missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Corning™ PYREX™ Tube Adapters with Outer standard taper Joint

The lower end is cut off at an angle for use as receiver adapter.

Supplier:  Corning™ 884024

Catalog No. 01-026-5


Explore available promotions

Description

Description

  • Made with inner angle of 105°
  • Upper end equipped with outer standard taper 24/40 joint
  • Tube O.D., 12mm

Order Info

Tube O.D., 12mm.

Specifications

Specifications

Tube Adapter
Borosilicate Glass (3.3)
24/40 Inner bottom, 24/40 outer top
105°
Videos
SDS
Documents

Documents

Promotions

Promotions