Corning™ PYREX™ VISTA™ Class B Volumetric Flasks with Polyethylene Snap-Cap
Capacity tolerances are twice those specified for Class A volumetric ware
Supplier: Corning™ 7058125
Description
These Class B volumetric flasks have capacity tolerances are twice those specified for Class A volumetric ware. The graduation line is sharp and permanent; large blue block letters make inscriptions easy to read. Snap caps are supplied with all sizes.
Specifications
To Contain | |
LDPE | |
Glass, Borosilicate Glass | |
12/Cs. | |
B | |
Snap Cap |
Blue | |
25 mL | |
Long | |
6/Pk. | |
Flat |