Corning™ PYREX™ VISTA™ Low Form Griffin Beaker
Provide balance between thermal shock resistance and mechanical strength
Supplier:  Corning™ 7000050
 View more versions of this product
Catalog No. 07-250-053
Explore available promotions
Description
Description
Provide balance between thermal shock resistance and mechanical strength
Corning™ PYREX™ VISTA™ Low Form Griffin Beakers provide balance between thermal shock resistance and mechanical strength.
These beakers with spout are manufactured to provide balance between thermal shock resistance and mechanical strength. For convenience, the 250 through 1000 mL beakers have a double graduated metric scale to indicate approximate content. All sizes have blue graduations and an extra large blue marking spot. The 10 mL size is not graduated. The beakers are designed to ASTM E-960.
Compliance
Designed to ASTM™ E 960 requirements.
Specifications
Specifications
1.69 oz.
4.2 cm
Borosilicate Glass
10 to 40 mL
5.6 cm
Extra Large Marking
Beaker
Yes
Blue Graduated Metric Scale
1.65 in.
50 mL
Provide Balance Between Thermal Shock Resistance and Mechanical Strength
2.2 in.
10 mL
24 Pk.
Reusable
ASTM E960
Griffin
Videos
SDS
Documents
Documents
Accessories