missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridazine-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3434103

 View more versions of this product

Catalog No. AAH3434103


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Pyridazine-3-carboxylic acid
∽195°C (decomposition)
MFCD01646333
Moisture sensitive
RUUOPSRRIKJHNH-UHFFFAOYSA-N
pyridazine-3-carboxylic acid
269369
97%
2164-61-6
C5H4N2O2
1 g
3-pyridazinecarboxylic acid, 3-carboxypyridazine, 3-pyridazine carboxylic acid, pubchem22109, pyridazine carboxylic acid, ksc202q0p, pyridazine-3-carboxylic acid, 3-carboxypyridazine; 3-pyridazinecarboxylic acid;
C1=CC(=NN=C1)C(=O)O
124.099
124.1

Chemical Identifiers

2164-61-6
124.099
RUUOPSRRIKJHNH-UHFFFAOYSA-N
269369
C1=CC(=NN=C1)C(=O)O
C5H4N2O2
MFCD01646333
3-pyridazinecarboxylic acid, 3-carboxypyridazine, 3-pyridazine carboxylic acid, pubchem22109, pyridazine carboxylic acid, ksc202q0p, pyridazine-3-carboxylic acid, 3-carboxypyridazine; 3-pyridazinecarboxylic acid;
pyridazine-3-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only