missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridazine-4-carboxylic Acid 98.0+%, TCI America™
$382.13 - $382.13
Chemical Identifiers
| CAS | 50681-25-9 |
|---|---|
| Molecular Formula | C5H4N2O2 |
| Molecular Weight (g/mol) | 124.099 |
| MDL Number | MFCD00011576 |
| InChI Key | JUSIWJONLKBPDU-UHFFFAOYSA-N |
| Synonym | 4-pyridazinecarboxylic acid, 4-carboxypyridazine, 4-pyridazinecarboxylicacid, pubchem9501, acmc-1alr8, pyridazin-4-carboxylic acid, 4-pyridazine-carboxylic acid, ksc269i2f, pyridazin-4-yl formate, 1,2-diazine-4-carboxylic acid |
| PubChem CID | 2761046 |
| IUPAC Name | pyridazine-4-carboxylic acid |
| SMILES | C1=CN=NC=C1C(=O)O |
Chemical Identifiers
| 50681-25-9 | |
| 124.099 | |
| JUSIWJONLKBPDU-UHFFFAOYSA-N | |
| 2761046 | |
| C1=CN=NC=C1C(=O)O |
| C5H4N2O2 | |
| MFCD00011576 | |
| 4-pyridazinecarboxylic acid, 4-carboxypyridazine, 4-pyridazinecarboxylicacid, pubchem9501, acmc-1alr8, pyridazin-4-carboxylic acid, 4-pyridazine-carboxylic acid, ksc269i2f, pyridazin-4-yl formate, 1,2-diazine-4-carboxylic acid | |
| pyridazine-4-carboxylic acid |
Specifications
| 50681-25-9 | |
| White | |
| MFCD00011576 | |
| 4-pyridazinecarboxylic acid, 4-carboxypyridazine, 4-pyridazinecarboxylicacid, pubchem9501, acmc-1alr8, pyridazin-4-carboxylic acid, 4-pyridazine-carboxylic acid, ksc269i2f, pyridazin-4-yl formate, 1,2-diazine-4-carboxylic acid | |
| C1=CN=NC=C1C(=O)O | |
| 124.099 | |
| 124.10 | |
| Crystalline Powder |
| 238°C | |
| C5H4N2O2 | |
| 1 g | |
| JUSIWJONLKBPDU-UHFFFAOYSA-N | |
| pyridazine-4-carboxylic acid | |
| 2761046 | |
| ≥98.0% (GC,T) | |
| Pyridazine-4-carboxylic Acid |
Safety and Handling
TSCA : No