missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridine-2,6-dicarboxylic acid, 98%, Thermo Scientific Chemicals

$67.26 - $513.79

Chemical Identifiers

CAS 499-83-2
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00006299
InChI Key WJJMNDUMQPNECX-UHFFFAOYSA-N
Synonym 2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid
PubChem CID 10367
ChEBI CHEBI:46837
IUPAC Name pyridine-2,6-dicarboxylic acid
SMILES C1=CC(=NC(=C1)C(=O)O)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1226314
View Documents
Thermo Scientific Chemicals
A1226314
25 g
Each for $67.26
 
AAA1226322
View Documents
Thermo Scientific Chemicals
A1226322
100 g
Each for $150.12
 
AAA1226336
View Documents
Thermo Scientific Chemicals
A1226336
500 g
Each for $513.79
 
Description

Description

Pyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyridine-2,6-dicarboxylic acid is used in the preparation of dipicolinato ligated lanthanide and transition metal complexes. It acts as a chelating agent for chromium, zinc, manganese, copper, iron and molybdenum. Its calcium-dipcolinic acid complex is used to protect deoxyribonucleic acid (DNA) from heat denaturation which enhances the DNA stability. It plays an important role as a marker for the effectiveness of sterilization.

Solubility
Insoluble in water.

Notes
Incompatible with acids, bases and oxidizing agents.
Specifications

Chemical Identifiers

499-83-2
167.12
WJJMNDUMQPNECX-UHFFFAOYSA-N
10367
pyridine-2,6-dicarboxylic acid
C7H5NO4
MFCD00006299
2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid
CHEBI:46837
C1=CC(=NC(=C1)C(=O)O)C(=O)O

Specifications

499-83-2
188°C (370°F)
MFCD00006299
131629
Insoluble in water.
C1=CC(=NC(=C1)C(=O)O)C(=O)O
167.12
CHEBI:46837
98%
∼245°C (decomposition)
C7H5NO4
25 g
2,6-pyridinedicarboxylic acid, dipicolinic acid, 2,6-dipicolinic acid, dipicolinate, 2,6-dicarboxypyridine, 2,6-pyridinedicarboxylate, unii-ue81s5cq0g, 2,6-pyridinedicarboxylicacid, ue81s5cq0g, 2,6-pyridinedicarboxylic acid dipicolinic acid
WJJMNDUMQPNECX-UHFFFAOYSA-N
pyridine-2,6-dicarboxylic acid
10367
167.12
Pyridine-2,6-dicarboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-894-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only