Learn More
Pyridine-2-carboxaldehyde, 99%, Thermo Scientific Chemicals
$68.70 - $454.95
Chemical Identifiers
CAS | 1121-60-4 |
---|---|
Molecular Formula | C6H5NO |
Molecular Weight (g/mol) | 107.112 |
MDL Number | MFCD00006290 |
InChI Key | CSDSSGBPEUDDEE-UHFFFAOYSA-N |
Synonym | 2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde |
PubChem CID | 14273 |
ChEBI | CHEBI:73012 |
IUPAC Name | pyridine-2-carbaldehyde |
SMILES | C1=CC=NC(=C1)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1139114
|
Thermo Scientific Chemicals
A1139114 |
25 g |
Each for $68.70
|
|
|||||
AAA1139122
|
Thermo Scientific Chemicals
A1139122 |
100 g |
Each for $139.79
|
|
|||||
AAA1139136
|
Thermo Scientific Chemicals
A1139136 |
500 g |
Each for $454.95
|
|
|||||
Description
Pyridine-2-carboxaldehyde, 99% is used as an Intermediate of Bisacodyl. It is also used as an intermediate in pharamceutical research, Intermediate in organicc synethesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyridine-2-carboxaldehyde, 99% is used as an Intermediate of Bisacodyl. It is also used as an intermediate in pharamceutical research, Intermediate in organicc synethesis.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature 2 - 8°C .Store under inert gas. Stable under recommended storage conditions.Incompatible to Strong bases, Strong oxidizing agents and acids.
Chemical Identifiers
1121-60-4 | |
107.112 | |
CSDSSGBPEUDDEE-UHFFFAOYSA-N | |
14273 | |
pyridine-2-carbaldehyde |
C6H5NO | |
MFCD00006290 | |
2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde | |
CHEBI:73012 | |
C1=CC=NC(=C1)C=O |
Specifications
1121-60-4 | |
1.124 | |
180°C to 181°C | |
Characteristic | |
1.536 | |
25 g | |
105341 | |
2-pyridinecarboxaldehyde, picolinaldehyde, pyridine-2-carboxaldehyde, 2-formylpyridine, pyridine-2-aldehyde, picolinal, 2-picolinaldehyde, 2-pyridaldehyde, picolinic aldehyde, 2-pyridylaldehyde | |
CSDSSGBPEUDDEE-UHFFFAOYSA-N | |
pyridine-2-carbaldehyde | |
14273 | |
107.11 | |
Pyridine-2-carboxaldehyde |
-21°C | |
6.5 | |
68°C (154°F) | |
C6H5NO | |
MFCD00006290 | |
UN2810 | |
Air and light sensitive | |
It is soluble in water. | |
C1=CC=NC(=C1)C=O | |
107.112 | |
CHEBI:73012 | |
99% |
Safety and Handling
GHS H Statement
H331-H318-H227-H302-H315-H317-H335
Toxic if inhaled.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P311-P330-P332+P313-P362-P370+P378q-P501c
H227-H302-H315-H318-H331
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 214-333-6
TSCA : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only