Learn More
Pyridine-3,4-dicarboxylic anhydride, 97%
CAS: 4664-08-8 | C7H3NO3 | 149.11 g/mol
$70.26 - $806.74
Chemical Identifiers
| CAS | 4664-08-8 |
|---|---|
| Molecular Formula | C7H3NO3 |
| Molecular Weight (g/mol) | 149.11 |
| MDL Number | MFCD00010680 |
| InChI Key | KFKMGUPDWTWQFM-UHFFFAOYSA-N |
| Synonym | furo 3,4-c pyridine-1,3-dione, 3,4-pyridinedicarboxylic anhydride, pyridine-3,4-dicarboxylic anhydride, 3,4-pyridinedicarboxylic acid anhydride, 1h,3h-furo 3,4-c pyridine-1,3-dione, cinchomeronic anhydride, pyridine-3,4-dicarboxylicanhydride, 1,3-dihydrofuro 3,4-c pyridine-1,3-dione, furano 3,4-c pyridine-1,3-dione, 3,4-pyridinecarboxylic acid anhydride |
| PubChem CID | 278398 |
| IUPAC Name | furo[3,4-c]pyridine-1,3-dione |
| SMILES | C1=CN=CC2=C1C(=O)OC2=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1077103
|
Thermo Scientific Chemicals
A1077103 |
1 g |
Each for $70.26
|
|
|||||
|
AAA1077106
|
Thermo Scientific Chemicals
A1077106 |
5 g |
Each for $192.42
|
|
|||||
|
AAA1077114
|
Thermo Scientific Chemicals
A1077114 |
25 g |
Each for $806.74
|
|
|||||
Description
Pyridine-3,4-dicarboxylic anhydride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyridine-3,4-dicarboxylic anhydride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Decomposes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture.
Chemical Identifiers
| 4664-08-8 | |
| 149.11 | |
| KFKMGUPDWTWQFM-UHFFFAOYSA-N | |
| 278398 | |
| C1=CN=CC2=C1C(=O)OC2=O |
| C7H3NO3 | |
| MFCD00010680 | |
| furo 3,4-c pyridine-1,3-dione, 3,4-pyridinedicarboxylic anhydride, pyridine-3,4-dicarboxylic anhydride, 3,4-pyridinedicarboxylic acid anhydride, 1h,3h-furo 3,4-c pyridine-1,3-dione, cinchomeronic anhydride, pyridine-3,4-dicarboxylicanhydride, 1,3-dihydrofuro 3,4-c pyridine-1,3-dione, furano 3,4-c pyridine-1,3-dione, 3,4-pyridinecarboxylic acid anhydride | |
| furo[3,4-c]pyridine-1,3-dione |
Specifications
| 4664-08-8 | |
| C7H3NO3 | |
| 1 g | |
| furo 3,4-c pyridine-1,3-dione, 3,4-pyridinedicarboxylic anhydride, pyridine-3,4-dicarboxylic anhydride, 3,4-pyridinedicarboxylic acid anhydride, 1h,3h-furo 3,4-c pyridine-1,3-dione, cinchomeronic anhydride, pyridine-3,4-dicarboxylicanhydride, 1,3-dihydrofuro 3,4-c pyridine-1,3-dione, furano 3,4-c pyridine-1,3-dione, 3,4-pyridinecarboxylic acid anhydride | |
| KFKMGUPDWTWQFM-UHFFFAOYSA-N | |
| furo[3,4-c]pyridine-1,3-dione | |
| 278398 | |
| 97% |
| 75°C to 79°C | |
| MFCD00010680 | |
| Moisture sensitive | |
| Decomposes in water. | |
| C1=CN=CC2=C1C(=O)OC2=O | |
| 149.11 | |
| 149.11 | |
| Pyridine-3,4-dicarboxylic anhydride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only