Learn More
Pyridine-3,5-dicarboxylic acid, 98%, Thermo Scientific Chemicals
$180.58 - $635.71
Chemical Identifiers
CAS | 499-81-0 |
---|---|
Molecular Formula | C7H5NO4 |
Molecular Weight (g/mol) | 167.12 |
MDL Number | MFCD00006393 |
InChI Key | MPFLRYZEEAQMLQ-UHFFFAOYSA-N |
Synonym | 3,5-pyridinedicarboxylic acid, dinicotinic acid, 5-carboxynicotinic acid, 3,5-pyridinedicarboxylicacid, 3,5-pyridindicarbons, 3,5-dipyridinecarboxylic acid, pubchem8057, acmc-209kjb, 5-carboxy nicotinic acid, pyridine carboxylate, 6f |
PubChem CID | 10366 |
ChEBI | CHEBI:46875 |
IUPAC Name | pyridine-3,5-dicarboxylic acid |
SMILES | OC(=O)C1=CC(=CN=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1154009
|
Thermo Scientific Chemicals
A1154009 |
10 g |
Each for $180.58
|
|
|||||
AAA1154018
|
Thermo Scientific Chemicals
A1154018 |
50 g |
Each for $635.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
499-81-0 | |
167.12 | |
MPFLRYZEEAQMLQ-UHFFFAOYSA-N | |
10366 | |
pyridine-3,5-dicarboxylic acid |
C7H5NO4 | |
MFCD00006393 | |
3,5-pyridinedicarboxylic acid, dinicotinic acid, 5-carboxynicotinic acid, 3,5-pyridinedicarboxylicacid, 3,5-pyridindicarbons, 3,5-dipyridinecarboxylic acid, pubchem8057, acmc-209kjb, 5-carboxy nicotinic acid, pyridine carboxylate, 6f | |
CHEBI:46875 | |
OC(=O)C1=CC(=CN=C1)C(O)=O |
Specifications
499-81-0 | |
C7H5NO4 | |
10 g | |
3,5-pyridinedicarboxylic acid, dinicotinic acid, 5-carboxynicotinic acid, 3,5-pyridinedicarboxylicacid, 3,5-pyridindicarbons, 3,5-dipyridinecarboxylic acid, pubchem8057, acmc-209kjb, 5-carboxy nicotinic acid, pyridine carboxylate, 6f | |
OC(=O)C1=CC(=CN=C1)C(O)=O | |
167.12 | |
CHEBI:46875 | |
98% |
∼325°C (decomposition) | |
MFCD00006393 | |
131640 | |
MPFLRYZEEAQMLQ-UHFFFAOYSA-N | |
pyridine-3,5-dicarboxylic acid | |
10366 | |
167.12 | |
Pyridine-3,5-dicarboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 207-893-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only