Learn More
Pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 451480050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pyridine-3-sulfonyl chloride hydrochloride | |
144.0°C to 145.0°C | |
Authentic | |
C5H4ClNO2S·ClH | |
pyridine-3-sulfonyl chloride hydrochloride, pyridine-3-sulfonyl chloride hcl, 3-pyridinesulfonyl chloride hydrochloride, pyridine-3-sulphonyl chloride hydrochloride, 3-pyridinesulfonyl chloride, hcl, 3-pyridinesulfonyl chloride, hydrochloride, 3-pyridylsulfonyl chloride hydrochloride, pyridine-3-sulfonylchloride hydrochloride, pyridin-3-ylsulfonyl chloride hydrochloride, 3-pyridylsulfonylchloride hcl | |
C1=CC(=CN=C1)S(=O)(=O)Cl.Cl | |
214.07 | |
214.07 |
42899-76-3 | |
26.37 to 29.74% (C) 2.21 to 2.50% (H) 6.15 to 6.94% (N) | |
94% min. as diethylsulfonamide (HPLC) | |
5 g | |
VIVPWOMJFLICOZ-UHFFFAOYSA-N | |
pyridine-3-sulfonyl chloride;hydrochloride | |
12571517 | |
95% |
Chemical Identifiers
42899-76-3 | |
214.07 | |
pyridine-3-sulfonyl chloride hydrochloride, pyridine-3-sulfonyl chloride hcl, 3-pyridinesulfonyl chloride hydrochloride, pyridine-3-sulphonyl chloride hydrochloride, 3-pyridinesulfonyl chloride, hcl, 3-pyridinesulfonyl chloride, hydrochloride, 3-pyridylsulfonyl chloride hydrochloride, pyridine-3-sulfonylchloride hydrochloride, pyridin-3-ylsulfonyl chloride hydrochloride, 3-pyridylsulfonylchloride hcl | |
pyridine-3-sulfonyl chloride;hydrochloride |
C5H4ClNO2S·ClH | |
VIVPWOMJFLICOZ-UHFFFAOYSA-N | |
12571517 | |
C1=CC(=CN=C1)S(=O)(=O)Cl.Cl |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair
GHS Signal Word: Danger
RUO – Research Use Only