missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

pyridine-3-sulfonyl chloride hydrochloride, 95%, Thermo Scientific™

$63.86 - $154.82

Chemical Identifiers

CAS 42899-76-3
Molecular Formula C5H5Cl2NO2S
Molecular Weight (g/mol) 214.06
InChI Key VIVPWOMJFLICOZ-UHFFFAOYSA-N
Synonym pyridine-3-sulfonyl chloride hydrochloride, pyridine-3-sulfonyl chloride hcl, 3-pyridinesulfonyl chloride hydrochloride, pyridine-3-sulphonyl chloride hydrochloride, 3-pyridinesulfonyl chloride, hcl, 3-pyridinesulfonyl chloride, hydrochloride, 3-pyridylsulfonyl chloride hydrochloride, pyridine-3-sulfonylchloride hydrochloride, pyridin-3-ylsulfonyl chloride hydrochloride, 3-pyridylsulfonylchloride hcl
PubChem CID 12571517
IUPAC Name pyridine-3-sulfonyl chloride;hydrochloride
SMILES C1=CC(=CN=C1)S(=O)(=O)Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC04103CB
SDS View Documents
Thermo Scientific Chemicals
CC04103CB
250 mg
Each for $63.86
 
CC04103DA
SDS View Documents
Thermo Scientific Chemicals
CC04103DA
1 g
Each for $154.82
 
Description

Description

Specifications

Chemical Identifiers

42899-76-3
214.06
pyridine-3-sulfonyl chloride hydrochloride, pyridine-3-sulfonyl chloride hcl, 3-pyridinesulfonyl chloride hydrochloride, pyridine-3-sulphonyl chloride hydrochloride, 3-pyridinesulfonyl chloride, hcl, 3-pyridinesulfonyl chloride, hydrochloride, 3-pyridylsulfonyl chloride hydrochloride, pyridine-3-sulfonylchloride hydrochloride, pyridin-3-ylsulfonyl chloride hydrochloride, 3-pyridylsulfonylchloride hcl
pyridine-3-sulfonyl chloride;hydrochloride
C5H5Cl2NO2S
VIVPWOMJFLICOZ-UHFFFAOYSA-N
12571517
C1=CC(=CN=C1)S(=O)(=O)Cl.Cl

Specifications

42899-76-3
C5H5Cl2NO2S
pyridine-3-sulfonyl chloride hydrochloride, pyridine-3-sulfonyl chloride hcl, 3-pyridinesulfonyl chloride hydrochloride, pyridine-3-sulphonyl chloride hydrochloride, 3-pyridinesulfonyl chloride, hcl, 3-pyridinesulfonyl chloride, hydrochloride, 3-pyridylsulfonyl chloride hydrochloride, pyridine-3-sulfonylchloride hydrochloride, pyridin-3-ylsulfonyl chloride hydrochloride, 3-pyridylsulfonylchloride hcl
C1=CC(=CN=C1)S(=O)(=O)Cl.Cl
214.06
214.07
Pyridine-3-sulfonyl chloride hydrochloride
95%
250 mg
VIVPWOMJFLICOZ-UHFFFAOYSA-N
pyridine-3-sulfonyl chloride;hydrochloride
12571517
95%
Videos
Safety and Handling

Safety and Handling

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'