missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridine-4-aldoxime 98.0+%, TCI America™

$173.79 - $345.62

Chemical Identifiers

CAS 696-54-8
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00006446
InChI Key OFYLBLSSPQTTHT-VMPITWQZSA-N
Synonym pyridine-4-aldoxime, 4-pyridinealdoxime, isonicotinaldehyde oxime, 4-pyridinecarboxaldehyde, oxime, isonicotinaldoxime, 4-hydroxyiminomethyl pyridine, 4-pyridinaldoxime, 4-pyridine aldoxime, pyridine, 4-formyl-, oxime, ccris 7744
PubChem CID 5324569
IUPAC Name (E)-N-[(pyridin-4-yl)methylidene]hydroxylamine
SMILES O\N=C\C1=CC=NC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P067310G
View Documents
TCI America
P067310G
10 g
Each for $173.79
 
P067325G
View Documents
TCI America
P067325G
25 g
Each for $345.62
 
Description

Description

Specifications

Chemical Identifiers

696-54-8
122.13
OFYLBLSSPQTTHT-VMPITWQZSA-N
5324569
O\N=C\C1=CC=NC=C1
C6H6N2O
MFCD00006446
pyridine-4-aldoxime, 4-pyridinealdoxime, isonicotinaldehyde oxime, 4-pyridinecarboxaldehyde, oxime, isonicotinaldoxime, 4-hydroxyiminomethyl pyridine, 4-pyridinaldoxime, 4-pyridine aldoxime, pyridine, 4-formyl-, oxime, ccris 7744
(E)-N-[(pyridin-4-yl)methylidene]hydroxylamine

Specifications

696-54-8
C6H6N2O
10 g
OFYLBLSSPQTTHT-VMPITWQZSA-N
(E)-N-[(pyridin-4-yl)methylidene]hydroxylamine
5324569
≥98.0% (T)
Pyridine-4-aldoxime
133°C
MFCD00006446
pyridine-4-aldoxime, 4-pyridinealdoxime, isonicotinaldehyde oxime, 4-pyridinecarboxaldehyde, oxime, isonicotinaldoxime, 4-hydroxyiminomethyl pyridine, 4-pyridinaldoxime, 4-pyridine aldoxime, pyridine, 4-formyl-, oxime, ccris 7744
O\N=C\C1=CC=NC=C1
122.13
122.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : NR9410000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'