missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyridine-4-boronic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC04212DA
Specifications
Pyridin-4-ylboronic acid | |
97% | |
C5H6BNO2 | |
pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid | |
B(C1=CC=NC=C1)(O)O | |
122.918 | |
122.92 |
1692-15-5 | |
Amber Glass Bottle | |
1g | |
QLULGIRFKAWHOJ-UHFFFAOYSA-N | |
pyridin-4-ylboronic acid | |
2734379 | |
97% |
Chemical Identifiers
1692-15-5 | |
122.918 | |
pyridine-4-boronic acid, 4-pyridineboronic acid, 4-pyridylboronic acid, pyridin-4-yl boronic acid, 4-pyridinylboronic acid, pyridin-4-yl-4-boronic acid, 4-pyridinebornic acid, pyridyl-4-boronic acid, 4-pyridinyl boronic acid, pyridine-4-ylboronic acid | |
pyridin-4-ylboronic acid |
C5H6BNO2 | |
QLULGIRFKAWHOJ-UHFFFAOYSA-N | |
2734379 | |
B(C1=CC=NC=C1)(O)O |