missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium bromide perbromide, 90+%, technical

CAS: 39416-48-3 | C5H6Br3N | 319.83 g/mol

$236.65 - $648.47

Chemical Identifiers

CAS 39416-48-3
Molecular Formula C5H6Br3N
Molecular Weight (g/mol) 319.83
MDL Number MFCD00013223
Synonym Pyridine hydrobromide, compound with bromine
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158361000
SDS View Documents
Thermo Scientific Chemicals
158361000
100 g Glass bottle
Each for $236.65
Only null left
 
AC158365000
SDS View Documents
Thermo Scientific Chemicals
158365000
500 g Glass bottle
Each for $648.47
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

39416-48-3
319.83
Pyridine hydrobromide, compound with bromine
C5H6Br3N
MFCD00013223

Specifications

39416-48-3
Brown to Orange-Red
90% min. (Iodometry)
C5H6Br3N
100 g
01,967; 05,568; 06,499; 09,399; 10,333; 14,268; 15,276
Pyridine hydrobromide, compound with bromine
319.83
90+%
Powder, Crystals or Chunks
127°C to 133°C
Authentic
Glass bottle
MFCD00013223
20, V,5, 181
15, 8085
Solubility in water: decomposes. Other solubilities: sparingly soluble in acetic acid
319.83
Technical
Pyridinium bromide perbromide, 95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 254-446-8

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only