missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridinium chlorochromate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 183671000

 View more versions of this product

Catalog No. AC183671000



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Pyridinium chlorochromate
26299-14-9
Orange
98%
C5H6ClCrNO3
100 g
15, 8086
Solubility in water: .. Other solubilities: insoluble in dichloromethane
215.56
Crystalline Powder
98%
205.0°C
Authentic
Plastic bottle
MFCD00013106
06,498; 07,308; 08,425; 09,397; 10,334; 11,450; 12,256; 13,237; 14,269; 15,57
PCC
215.56
98%

Chemical Identifiers

26299-14-9
215.56
PCC
C5H6ClCrNO3
MFCD00013106
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause cancer by inhalation.
May intensify fire; oxidizer.

GHS P Statement:
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 247-595-5

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only