Learn More
Pyridoxal-5-phosphate hydrate, 98%, Thermo Scientific Chemicals
$74.95 - $723.48
Chemical Identifiers
CAS | 41468-25-1 |
---|---|
Molecular Formula | C8H10NO6P |
Molecular Weight (g/mol) | 247.14 |
MDL Number | MFCD00006333 |
InChI Key | NGVDGCNFYWLIFO-UHFFFAOYSA-N |
Synonym | 4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate |
PubChem CID | 38882 |
IUPAC Name | (4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate;hydrate |
SMILES | CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1232303
|
Thermo Scientific Chemicals
A1232303 |
1 g |
Each for $74.95
|
|
|||||
AAA1232306
|
Thermo Scientific Chemicals
A1232306 |
5 g |
Each for $191.92
|
|
|||||
AAA1232314
|
Thermo Scientific Chemicals
A1232314 |
25 g |
Each for $723.48
|
|
|||||
Description
Active form of Vitamin B-6Pyridoxal-5â-phosphate monohydrate is used to modify the lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions and as an inhibitor for purinergic receptors and intracellular. It is involved in decarboxylation and deamination reactions viz. the conversion of dopa into dopamine of glutamate into GABA and histidine to histamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsActive form of Vitamin B-6Pyridoxal-5â-phosphate monohydrate is used to modify the lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions and as an inhibitor for purinergic receptors and intracellular. It is involved in decarboxylation and deamination reactions viz. the conversion of dopa into dopamine of glutamate into GABA and histidine to histamine.
Solubility
Soluble in alkaline solutions. Sparingly soluble in water.
Notes
Light sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
Chemical Identifiers
41468-25-1 | |
247.14 | |
NGVDGCNFYWLIFO-UHFFFAOYSA-N | |
38882 | |
CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O |
C8H10NO6P | |
MFCD00006333 | |
4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate | |
(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate;hydrate |
Specifications
41468-25-1 | |
C8H10NO6P | |
1 g | |
14,7979 | |
Soluble in alkaline solutions. Sparingly soluble in water. | |
CC1=NC=C(COP(O)(O)=O)C(C=O)=C1O | |
247.14 | |
265.17 (247.15 Anhydrous) | |
Pyridoxal-5-phosphate hydrate |
142°C | |
MFCD00006333 | |
Light sensitive | |
4-formyl-5-hydroxy-6-methylpyridin-3-yl methyl dihydrogen phosphate hydrate, pyridoxal phosphate hydrate, pyridoxal 5-phosphate monohydrate, phosphopyridoxal monohydrate, pyridoxyl phosphate monohydrate, unii-5v5ioj8338, pyridoxal-5-phosphate, monohydrate, pyridoxal-5'-phosphate monohydrate, pyridoxal-5-phosphate monohydrate, pyridoxal 5'-phosphate monohydrate | |
NGVDGCNFYWLIFO-UHFFFAOYSA-N | |
(4-formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate;hydrate | |
38882 | |
98% |
Safety and Handling
EINECSNumber : 200-208-3
RTECSNumber : UV1207000
TSCA : Yes
Recommended Storage : Store at -20°C; Store under Nitrogen
RUO – Research Use Only