missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyridoxine hydrochloride, 98%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210277710

 View more versions of this product

Catalog No. ICN10277710


Explore available promotions

Description

Description

Specifications

Specifications

Pyridoxine hydrochloride
White
>98%
MFCD00012807
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
CC1=NC=C(C(=C1O)CO)CO.Cl
205.638
CHEBI:30961
0.98
58-56-0
1.43g/mL
C8H12ClNO3
10 g
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
6019
205.64
Powder

Chemical Identifiers

58-56-0
205.638
ZUFQODAHGAHPFQ-UHFFFAOYSA-N
6019
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride
C8H12ClNO3
MFCD00012807
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion
CHEBI:30961
CC1=NC=C(C(=C1O)CO)CO.Cl
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-386-2

Recommended Storage : Store at Room Temperature.

SDS
Documents

Documents

Promotions

Promotions