missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyridoxine hydrochloride, 98%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210277750
Specifications
Pyridoxine hydrochloride | |
White | |
>98% | |
MFCD00012807 | |
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion | |
CC1=NC=C(C(=C1O)CO)CO.Cl | |
205.638 | |
CHEBI:30961 | |
0.98 |
58-56-0 | |
1.43g/mL | |
C8H12ClNO3 | |
50 g | |
ZUFQODAHGAHPFQ-UHFFFAOYSA-N | |
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride | |
6019 | |
205.64 | |
Powder |
Chemical Identifiers
58-56-0 | |
205.638 | |
ZUFQODAHGAHPFQ-UHFFFAOYSA-N | |
6019 | |
4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol;hydrochloride |
C8H12ClNO3 | |
MFCD00012807 | |
pyridoxine hydrochloride, pyridoxine hcl, pyridoxol hydrochloride, vitamin b6, alestrol, becilan, benadon, hexavibex, hexermin, hexobion | |
CHEBI:30961 | |
CC1=NC=C(C(=C1O)CO)CO.Cl |
Safety and Handling
EINECSNumber : 200-386-2
Recommended Storage : Store at Room Temperature.