Learn More
Pyrite, naturally occurring mineral, grains, approximately 1.5-4.8mm (0.06-0.19in), Thermo Scientific Chemicals
For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals
Supplier: Thermo Scientific Chemicals 04263303
Description
For grain mounts, XRD and microprobe standards, identification of unknown minerals.Pyrite is used in the preparation of iron(II) sulfate, sulfur dioxide and iron(II) sulfide. It finds application as semiconductor material, as the cathode material in energizer band and non-rechargeable lithium batteries. It is utilized to make marcasite jewelry. In addition, it is used in photovoltaic solar panels. It is useful for grain mounts, XRD and microprobe standards and identification of unknown minerals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Naturally occurring mineral; Hardness: 6-6.5Specifications
Pyrite | |
Grains or Chunks | |
FeS2 | |
iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in | |
NFMAZVUSKIJEIH-UHFFFAOYSA-N | |
bis(sulfanylidene)iron | |
14788 | |
3.08 | |
5.02 |
1309-36-0 | |
1 g | |
MFCD00064690 | |
Insoluble in water. | |
S=[Fe]=S | |
119.965 | |
119.98 | |
Grains/chunks |
Chemical Identifiers
1309-36-0 | |
119.965 | |
NFMAZVUSKIJEIH-UHFFFAOYSA-N | |
14788 | |
S=[Fe]=S |
FeS2 | |
MFCD00064690 | |
iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in | |
bis(sulfanylidene)iron |
Safety and Handling
EINECSNumber : 215-167-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only