missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrite, naturally occurring mineral, grains, approximately 1.5-4.8mm (0.06-0.19in), Thermo Scientific Chemicals

For Grain Mounts, XRD and Microprobe Standards, Identification of Unknown Minerals

Supplier:  Thermo Scientific Chemicals 04263314

 View more versions of this product

Catalog No. AA4263314

Explore available promotions

Description

Description

For grain mounts, XRD and microprobe standards, identification of unknown minerals.Pyrite is used in the preparation of iron(II) sulfate, sulfur dioxide and iron(II) sulfide. It finds application as semiconductor material, as the cathode material in energizer band and non-rechargeable lithium batteries. It is utilized to make marcasite jewelry. In addition, it is used in photovoltaic solar panels. It is useful for grain mounts, XRD and microprobe standards and identification of unknown minerals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Naturally occurring mineral; Hardness: 6-6.5
Specifications

Specifications

Pyrite
Grains or Chunks
FeS2
iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in
NFMAZVUSKIJEIH-UHFFFAOYSA-N
bis(sulfanylidene)iron
14788
3.08
5.02 g/cm3 at 20°C
1309-36-0
25 g
MFCD00064690
Insoluble in water.
S=[Fe]=S
119.965
119.98
Grains/chunks

Chemical Identifiers

1309-36-0
119.965
NFMAZVUSKIJEIH-UHFFFAOYSA-N
14788
S=[Fe]=S
FeS2
MFCD00064690
iron disulfide, iron sulfide, pyrite, bis sulfanylidene iron, disulfanylideneiron, iron disulfide iron pyrite, marcasite, naturally occuring mineral, grains, approximately 0.06-0.19 in, pyrite, naturally occurring mineral, grains, approximately 0.06-0.19in
bis(sulfanylidene)iron
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-167-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only